Learn More
USA Scientific Inc Bead Genie, 120V
Catalog No. 502745502
Encompass
Bead Genie Bead beater. Spacing-saving design easily fits on the lab bench and is ideal for bead-beating procedures, utilizing different types of beads for lysing, extraction, or homogenization applications. Supplied with 4 Tube Holders - 12 x 1.5 / 2.0 mL 9 vertical & horizontal), 6 x15mL, and 3 x 50mL.
Catalog No. 50-274-5502 Supplier USA Scientific Inc Supplier No. 74031000
Bead Genie Bead beater. Spacing-saving design easily fits on the lab bench and is ideal for bead-beating procedures, utilizing different types of beads for lysing, extraction, or homogenization applications. Supplied with 4 Tube Holders - 12 x 1.5 / 2.0 mL 9 vertical & horizontal), 6 x15mL, and 3 x 50mL.