Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Bead Genie

Catalog No. 01804391 Shop All OHAUS Products
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-804-391 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-804-391 Supplier OHAUS Supplier No. 83041308
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bead beater ideal for lysing, extraction or homogenization applications, utilizing an aggressive vertical mixing motion.

  • Supplied with holders to hold vertically twelve microtubes (1.5 or 2.0 mL), six 15 mL tubes or three 50 mL tubes, and twelve microtubes (1.5 or 2.0 mL) horizontally.
  • Beads available and sold separately for complete bead beating solution.
  • The Bead Genie is compact compared to most bead beaters and homogenizers.
TRUSTED_SUSTAINABILITY

Specifications

Orbit 19 mm
Speed Range 250 to 2400 rpm
Voltage 120 V
Product Type Bead Beater
Model Bead Genie
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.