Learn More
Research Products International Corp Bead Genie Bead Beater
SDP
Catalog No. 502114064
Encompass
The Bead Genie is ideal for bead beating, lysing, extraction,or homogenization applications. Compact,space saving design, easily fits on the lab bench, into a bio safety cabinet, or for use in your cold room. High efficiency DC motor provides powerful, unique vertical motion, to ensure smooth and efficient operation.
The Bead Genie helps to facilitate many sample preparation requirements for Clinical labs, Research labs, Environmental and Agricultural research and testing laboratories. Available with a variety of accessories to fit common sample tubes. Supplied with 4 tube holders that accept 12 x 1.5/2.0ml (vertical & horizontal), 6 x 15 ml and 3 x 50mlsize tubes.
Powerful motor
Compact, efficient design
Variety of tube accessories
Catalog No. 50-211-4064 Supplier Research Products International Corp Supplier No. 155566
The Bead Genie is ideal for bead beating, lysing, extraction,or homogenization applications. Compact,space saving design, easily fits on the lab bench, into a bio safety cabinet, or for use in your cold room. High efficiency DC motor provides powerful, unique vertical motion, to ensure smooth and efficient operation.
The Bead Genie helps to facilitate many sample preparation requirements for Clinical labs, Research labs, Environmental and Agricultural research and testing laboratories. Available with a variety of accessories to fit common sample tubes. Supplied with 4 tube holders that accept 12 x 1.5/2.0ml (vertical & horizontal), 6 x 15 ml and 3 x 50mlsize tubes.
Powerful motor
Compact, efficient design
Variety of tube accessories