Learn More
Grainger Baileigh Industrial Bead Roller, 16 Gauge Steel, 36 in. Throat, 110 V, 1 ea
Catalog No. 502788014
Encompass_Preferred
The powered bead rolling machine is a robust tool designed for precision forming of large sheet-metal surfaces. Featuring a 36 in. throat depth and a rigid, fully welded steel frame, this machine is ideal for automotive hoods, deck lids, HVAC panels, and aircraft skins. It is powered by 110 V and utilizes a variable-speed foot pedal for controlled, hands-free operation.
  • Capable of forming up to 16-gauge mild steel and 20-gauge stainless steel.
  • Includes a variable-speed foot pedal for adjustable rolling speed.
  • Operates in both forward and reverse directions.
  • Top roll adjustment via lead screw and handle allows for consistent bead depth.
  • Equipped with bronze bushings at all rotating points for durability.
  • Supplied with a floor stand featuring a storage rack for up to 10 tooling sets.
  • Backed by a 1-year parts warranty.
Catalog No. 50-278-8014 Supplier Grainger Supplier No. 18F321
The powered bead rolling machine is a robust tool designed for precision forming of large sheet-metal surfaces. Featuring a 36 in. throat depth and a rigid, fully welded steel frame, this machine is ideal for automotive hoods, deck lids, HVAC panels, and aircraft skins. It is powered by 110 V and utilizes a variable-speed foot pedal for controlled, hands-free operation.
  • Capable of forming up to 16-gauge mild steel and 20-gauge stainless steel.
  • Includes a variable-speed foot pedal for adjustable rolling speed.
  • Operates in both forward and reverse directions.
  • Top roll adjustment via lead screw and handle allows for consistent bead depth.
  • Equipped with bronze bushings at all rotating points for durability.
  • Supplied with a floor stand featuring a storage rack for up to 10 tooling sets.
  • Backed by a 1-year parts warranty.