Description
- Reduced mass allowing higher sample size
- Dry or wet grinding
- Durable and easy to clean
- Process samples in up to eight 50 mL tubes
Specifications
Specifications
| Autoclavable | Autoclavable |
| Blade Material | Stainless Steel |
| For Use With (Application) | Dry or Wet Grinding |
| For Use With (Equipment) | Omni International Bead Ruptor 96 |
| Holds | 8 x 50 mL Tubes |
| Includes | Tube Holder Set (Left and Right) |
| Product Type | Rack Accessory |