Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omni International Bead Ruptor 96 Microtube or Screw Cap Tube Holders
GREENER_CHOICE

Catalog No. 15341109 Shop All Omni International Inc Products
Change view
Click to view available options
For Use With (Equipment):
Bead Ruptor 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
15-341-109 Bead Ruptor 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-341-109 Supplier Omni International Supplier No. 27106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bead Ruptor 96 Microtube or Screw Cap Tube Holders hold microcentrifuge or screw cap tubes securely in place during processing.

  • Includes a pair of two PTFE tube adapters
  • Each pair will hold 24 x 0.5 mL screw cap tubes, 24 x 2 mL screw cap tubes, 24 x 1.5 mL microcentrifuge tubes or 24 x 2 mL microcentrifuge tubes.

Specifications

Product Type Tube Holder
Sample Volume (Metric) 200 μL to 1.5 mL
Autoclavable Not Autoclavable
Color White
For Use With (Equipment) Bead Ruptor 96
Holds 24 x 0.5 mL screw cap tubes, 24 x 2 mL screw cap tubes, 24 x 1.5 mL microcentrifuge tubes, or 24 x 2 mL microcentrifuge tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.