Learn More
Omni International Bead Ruptor 96 Stainless Steel Milling Balls
GREENER_CHOICE
Catalog No. 15341116 Shop All Omni International Inc Products
Change view
Click to view available options
Diameter (Metric):
10 mm
25 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
15-341-116 10 mm
15-341-117 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-341-116 Supplier Omni International Supplier No. 27203
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stainless Steel Milling Balls must be used with stainless steel jars.
  • Set of Stainless Steel Milling Balls
Specifications
Product Type Milling Balls
Diameter (Metric) 10 mm
Autoclavable Autoclavable
Material Stainless Steel
For Use With (Application) Dry Grinding or Cryomilling
For Use With (Equipment) Stainless Steel Jars
Packaging Quantity 10 Pk.