Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omni International Bead Ruptor Cryo Cooling Unit
GREENER_CHOICE

Catalog No. 17100870 Shop All Omni International Inc Products
Change view
Click to view available options
Capacity (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
17-100-870 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-870 Supplier Omni International Supplier No. 198005
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This cooling system protects heat-sensitive samples during homogenization.

  • Compatible with the Bead Ruptor Elite
  • Designed to prevent the increase of sample temperature during the homogenization process
  • The friction generated through the bead milling process generates heat that is imparted to the sample
  • In order to maintain a constant processing temperature during homogenization, the BR Cryo passes chilled air (-100°C to -50°C) into the sample processing chamber and across the sample tubes to dissipate heat
  • Samples can be maintained at a temperature ranging from 4°C to ambient, thus protecting heat sensitive samples such as RNA and proteins
  • Can be used to maintain a constant temperature when extracting functional enzymes or volatile small molecules
  • Warranty: 1 year
  • Note: Not compatible with Tube Carriages >7 mL or the 48 Position 2 mL Carriage

Specifications

Speed 7 m/s
Capacity (Metric) 1 L
Certifications/Compliance CE, UKCA
Weight (English) 16 lb.
Weight (Metric) 7.2 kg
Product Type Bead Mill Homogenizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.