Learn More
Macherey-Nagel MN Bead Tubes Type G, 50/pk
Supplier:  Macherey-Nagel 740817.50
Encompass_Preferred
2 mL tubes with 5 mm steel beads; for homogenization of plant material.
Catalog No. NC1883495