Learn More
Cole-Parmer Extech TP870 Thermocouple Probe, Beaded Wire, Type K
Supplier:  Cole-Parmer 9041510
Encompass
Type K temperature probe with mini connector. Temperature range:-40 to 482-Degrees F (-40 to 250-Degrees C). Type K mini connector. Compatible with most meters that have Type K temperature function.
Catalog No. 50-993-114