Learn More
Alkali Scientific BeadBath Duo 2L Dual Function
SDP
Catalog No. 503029491
Encompass_Preferred
The BeadBath Duo series are the first laboratory baths designed for precision temperature control when used with water or with (the increasingly popular) Lab Armor thermal beads Unlike any other laboratory bath they feature a MODE button that can be used to toggle between Bead or Water mode Choosing the correct mode will automatically adjust the operation and balance of the multiple internal heating and sensing elements to ensure optimal performance The result is one combination bath that is suitable for accurate and uniform temperature control in either condition While the majority of research laboratories today are using water baths many prefer the convenience of bead baths due to the advantages listed below With the BeadBath Duo the lab gets the best of both worlds allowing conversion from one bath type to the other with the simple press of a button Two separate modes for BEADS or WATER Quik-Cal simple in-lab calibration function Offers all of the advantages of a Bead Bath El
Catalog No. 50-302-9491 Supplier Alkali Scientific Supplier No. WB709
The BeadBath Duo series are the first laboratory baths designed for precision temperature control when used with water or with (the increasingly popular) Lab Armor thermal beads Unlike any other laboratory bath they feature a MODE button that can be used to toggle between Bead or Water mode Choosing the correct mode will automatically adjust the operation and balance of the multiple internal heating and sensing elements to ensure optimal performance The result is one combination bath that is suitable for accurate and uniform temperature control in either condition While the majority of research laboratories today are using water baths many prefer the convenience of bead baths due to the advantages listed below With the BeadBath Duo the lab gets the best of both worlds allowing conversion from one bath type to the other with the simple press of a button Two separate modes for BEADS or WATER Quik-Cal simple in-lab calibration function Offers all of the advantages of a Bead Bath El