Learn More
Research Products International Corp BeadBeater Ice Water Jacket
SDP
Catalog No. 501978015
Encompass
For use with the Beadbeater to keep temperature increases under control during mechanical cell disruption.
Catalog No. 50-197-8015 Supplier Research Products International Corp Supplier No. 141817
For use with the Beadbeater to keep temperature increases under control during mechanical cell disruption.