Learn More
Research Products International Corp Bead Beater, Small Stainless Steel Chamber, 50 ml Working Volume
SDP
Catalog No. 50550168
Encompass
These chambers have working volumes (i.e., cells plus buffer) of 50 ml and a maximum sample size of 12g. The chambers are made of stainless steel and polypropylene plastic and can be autoclaved, if desired. The small chambers fit inside the standard 350 ml BeadBeater polycarbonate chamber. The latter may be filled with crushed ice and water and to help keep the sample cold.
Catalog No. 50-550-168 Supplier Research Products International Corp Supplier No. 141816
These chambers have working volumes (i.e., cells plus buffer) of 50 ml and a maximum sample size of 12g. The chambers are made of stainless steel and polypropylene plastic and can be autoclaved, if desired. The small chambers fit inside the standard 350 ml BeadBeater polycarbonate chamber. The latter may be filled with crushed ice and water and to help keep the sample cold.