Learn More
Research Products International Corp BeadBeater, Small Stainless Steel Chamber, 15 ml Working Volume
SDP
Catalog No. 501977325
Encompass
Chambers have working volumes (i.e., cells plus buffer) of 15 ml and a maximum sample size of 7g. The chambers are made of stainless steel and polypropylene plastic and can be autoclaved, if desired. The small chambers fit inside the standard 350 ml BeadBeater polycarbonate chamber. The latter may be filled with crushed ice and water and to help keep the sample cold.
Catalog No. 50-197-7325 Supplier Research Products International Corp Supplier No. 141815
Chambers have working volumes (i.e., cells plus buffer) of 15 ml and a maximum sample size of 7g. The chambers are made of stainless steel and polypropylene plastic and can be autoclaved, if desired. The small chambers fit inside the standard 350 ml BeadBeater polycarbonate chamber. The latter may be filled with crushed ice and water and to help keep the sample cold.