Learn More
BioSpec Products BeadBeater for 0.5 mm glass beads
SDP
Catalog No. NC0440112
Encompass_Preferred
The BeadBeater is the ultimate laboratory-scale homogenizer for disrupting aqueous suspensions of yeast, fungi or bacteria. Complete cell lysis of 10-80 g (wet weight) of cells is achieved in about three minutes of operation. The BeadBeater comes complete with a 350 ml polycarbonate chamber, rotor assembly, motor base, ice-water cooling jacket, and one pound of 0.5 mm glass beads.
Catalog No. NC0440112 Supplier BioSpec Products Supplier No. 1107900105
The BeadBeater is the ultimate laboratory-scale homogenizer for disrupting aqueous suspensions of yeast, fungi or bacteria. Complete cell lysis of 10-80 g (wet weight) of cells is achieved in about three minutes of operation. The BeadBeater comes complete with a 350 ml polycarbonate chamber, rotor assembly, motor base, ice-water cooling jacket, and one pound of 0.5 mm glass beads.