Learn More
Research Products International Corp BEADBLASTER 24R REFRIGERATED
SDP
Supplier:  Research Products International Corp 985386R
Encompass
The Beadblaster 24R is a highly efficient bead homogenizer with built in refrigeration to protect temperature sensitive molecules from heat degradation and denaturation. Cooling function protects temperature sensitive samples and provides true refrigeration - no dry ice, liquid nitrogen or external connections.
Catalog No. NC1873483
Add to Cart
Request a Quote