Learn More
Alkali Scientific Beadblaster 96 Ball Mill Homog
SDP
Catalog No. 503029655
Encompass_Preferred
The BeadBlaster96 is an extremely versatile bead mill homogenizer that has applications in a variety of areas including biological research environmental testing and industrial settings Adapters are available for microplates microtubes and 15/50ml tubes Grinding jars and balls in a variety of materials are available Wet homogenization dry grinding and cryogrinding can all be performed with the BeadBlaster96 The arced motion of the sample holders and powerful motor allow samples to be processed quickly and efficiently All parameters are digitally controlled through the large touch screen and 9 programs can be stored in memory The safety cover lifts up for easy access to the sample holders and automatically locks during operation The wrap-around design of the clear acrylic lid allows for visualization during homogenization Wet dry or cryogrinding Accepts a variety of tubes plates and jars Large digital touch screen control panel See-though safety lid
Catalog No. 50-302-9655 Supplier Alkali Scientific Supplier No. RS2053
Add to Cart
Add to Cart
Request a Quote
The BeadBlaster96 is an extremely versatile bead mill homogenizer that has applications in a variety of areas including biological research environmental testing and industrial settings Adapters are available for microplates microtubes and 15/50ml tubes Grinding jars and balls in a variety of materials are available Wet homogenization dry grinding and cryogrinding can all be performed with the BeadBlaster96 The arced motion of the sample holders and powerful motor allow samples to be processed quickly and efficiently All parameters are digitally controlled through the large touch screen and 9 programs can be stored in memory The safety cover lifts up for easy access to the sample holders and automatically locks during operation The wrap-around design of the clear acrylic lid allows for visualization during homogenization Wet dry or cryogrinding Accepts a variety of tubes plates and jars Large digital touch screen control panel See-though safety lid