Learn More
Sigma Aldrich Fine Chemicals Biosciences BeadBlasterTM 24 Microtube1EA
Catalog No. 502908791
Encompass
The BeadBlaste 24 completely lyses grinds and homogenizes a broad variety of biological samples. Even the most difficult and resistant samples are fully homogenized often in 35 seconds or less..nbspFor all sample types from soft tissue to bone.nbspMix up to 24 tubes simultaneously.nbspSealed 2.0ml tubes - eliminates cross contamination
Catalog No. 50-290-8791 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424751EA
Add to Cart
Add to Cart
Request a Quote
The BeadBlaste 24 completely lyses grinds and homogenizes a broad variety of biological samples. Even the most difficult and resistant samples are fully homogenized often in 35 seconds or less..nbspFor all sample types from soft tissue to bone.nbspMix up to 24 tubes simultaneously.nbspSealed 2.0ml tubes - eliminates cross contamination