Learn More
Alkali Scientific BeadBug™ Three Position Microtube Homogenizer, 115V
SDP
Catalog No. 502336157
Encompass_Preferred
The BeadBug™ is a high energy benchtop homogenizer that sets the performance standard for personal sized cell disruption and lysis instruments. Simultaneous homogenization of up to three samples takes place (often within 45 seconds) inside disposable 2mL screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type. Developed for samples too tough to be homogenized in a common lab vortexer, the BeadBug™ provides a convenient and economical alternative to the traditional, over-priced homogenizers on the market today. With up to three tube capacity and a foot print of just 7 X 8", the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.
Catalog No. 50-233-6157 Supplier Alkali Scientific Supplier No. RS7020
The BeadBug™ is a high energy benchtop homogenizer that sets the performance standard for personal sized cell disruption and lysis instruments. Simultaneous homogenization of up to three samples takes place (often within 45 seconds) inside disposable 2mL screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type. Developed for samples too tough to be homogenized in a common lab vortexer, the BeadBug™ provides a convenient and economical alternative to the traditional, over-priced homogenizers on the market today. With up to three tube capacity and a foot print of just 7 X 8", the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.