Learn More
Research Products International Corp BeadBug MicroTube Homogenizer
SDP
Catalog No. 501367807
Encompass
BeadBug 3 is a powerful benchtop homogenizer for performing cell lysis, tissue grinding and sample homogenizing. Easy to use, simply place sample and disruption beads into a standard 2.0ml screw cap micro-tube and insert tube in holder. BeadBug will process up to three samples at one time with complete homogenizing within a few minutes or less. Heavy duty mixing chamber produces a sample disruption using hardened glass or zirconium beads.
Compact unit has a small footprint of 7 x 8 Inches for convenient use on crowded benchtops.
Catalog No. 50-136-7807 Supplier Research Products International Corp Supplier No. 985385
BeadBug 3 is a powerful benchtop homogenizer for performing cell lysis, tissue grinding and sample homogenizing. Easy to use, simply place sample and disruption beads into a standard 2.0ml screw cap micro-tube and insert tube in holder. BeadBug will process up to three samples at one time with complete homogenizing within a few minutes or less. Heavy duty mixing chamber produces a sample disruption using hardened glass or zirconium beads.
Compact unit has a small footprint of 7 x 8 Inches for convenient use on crowded benchtops.