Learn More
Sigma Aldrich Fine Chemicals Biosciences BeadBugTM 6 microtube homo1EA
Catalog No. 502908765
Encompass
Like the other members of the Benchmark Microtube Homogenizer family the BeadBug 6 utilizes a three dimensional shaking motion to disrupt samples. This motion when combined with beads of the appropriate size and material causes high velocity impacts with the sample and is an extremely fast and efficient method for releasing cellular contents including nucleic acids and proteins.Many different sample types from cells in culture to organ tissues and bone can be homogenized in the BeadBug 6 most within 45 seconds. Programming the unit is very simple - just set the speed time number of cycles and pausebetween cycles then press start. For convenience the last programmed parameters remain in memory for repeated runs. Cycle times can be set between 1 and 90 seconds with pauses between cycles up to 90 seconds in 1 second increments. A maximun of 10 cycles can be set per program.Sealed tubes prevent cross contamination in the BeadBug 6. Samples are simply loaded into the tubes along w
Catalog No. 50-290-8765 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7426821EA
Like the other members of the Benchmark Microtube Homogenizer family the BeadBug 6 utilizes a three dimensional shaking motion to disrupt samples. This motion when combined with beads of the appropriate size and material causes high velocity impacts with the sample and is an extremely fast and efficient method for releasing cellular contents including nucleic acids and proteins.Many different sample types from cells in culture to organ tissues and bone can be homogenized in the BeadBug 6 most within 45 seconds. Programming the unit is very simple - just set the speed time number of cycles and pausebetween cycles then press start. For convenience the last programmed parameters remain in memory for repeated runs. Cycle times can be set between 1 and 90 seconds with pauses between cycles up to 90 seconds in 1 second increments. A maximun of 10 cycles can be set per program.Sealed tubes prevent cross contamination in the BeadBug 6. Samples are simply loaded into the tubes along w