Learn More
Sigma Aldrich Fine Chemicals Biosciences BeadBugTM microtube homoge1EA
Catalog No. 502908604
Encompass
The BeadBug(TM) is a high energy benchtop homogenizer that sets the performance standard for personal sized cell disruption and lysis instruments.Simultaneous homogenization of up to 3 samples takes place (often within 45 seconds) inside the disposable 2 mL screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type.With up to three tube capacity and a foot print of just 7 x 8 in. the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.
Catalog No. 50-290-8604 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7637051EA
The BeadBug(TM) is a high energy benchtop homogenizer that sets the performance standard for personal sized cell disruption and lysis instruments.Simultaneous homogenization of up to 3 samples takes place (often within 45 seconds) inside the disposable 2 mL screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type.With up to three tube capacity and a foot print of just 7 x 8 in. the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.