Learn More
Sigma Aldrich Fine Chemicals Biosciences BeadBugTM prefilled tubes
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z76374850EA
Encompass
These prefilled tubes fit the BeadBug(TM) microtube homogenizer and come with standard acid-washed glass beads triple-pure zirconium beads or stainless steel beads.The triple-pure beads are molecular biology grade and are acid washed heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well.All tubes are non-skirted with screw-caps
Catalog No. 50-290-1260