Learn More
Ace Glass, Inc. Coupling, makes bead to bead coupling of 1 inch glass process drain pipe, 35in-lbs max torque, max temp400°F (204°C)
SDP
Supplier:  Ace Glass, Inc. 885607
Encompass
Catalog No. NC2093282