Learn More
Chemglass Life Sciences Beads, Borosilicate Glass, 4mm, Approx. 5, 500 Pcs. Per Lb.
SDP
Catalog No. 50872931
Encompass_Preferred
Beads, Borosilicate Glass, 4mm, Approx. 5,500 Pcs. Per Lb. Solid borosilicate spheres can be used as distillation column packing, boiling stones or to provide support for chromatography columns used in gel filtrations. Beads have excellent chemical resistance. Packed in boxes containing one pound.
Catalog No. 50-872-931 Supplier Chemglass Life Sciences Supplier No. CG110103
Beads, Borosilicate Glass, 4mm, Approx. 5,500 Pcs. Per Lb. Solid borosilicate spheres can be used as distillation column packing, boiling stones or to provide support for chromatography columns used in gel filtrations. Beads have excellent chemical resistance. Packed in boxes containing one pound.