Learn More
Med Vet International JorVet GI Magic Beads, Small Pack, 5 Doses, J1146
Catalog No. 5000495722
Encompass_Preferred
Single Dose: 4 small capsules: with 30 x 1.5mm beads x 10 x 4 .7mm beads 5 doses per box JorVet gI Magic Beads. Is that a GI foreign body or not? Liquid Barium is useful for diagnosing suspected GI foreign bodies on radiographs.
Catalog No. 50-004-95722 Supplier Med Vet International Supplier No. JOR1146
Single Dose: 4 small capsules: with 30 x 1.5mm beads x 10 x 4 .7mm beads 5 doses per box JorVet gI Magic Beads. Is that a GI foreign body or not? Liquid Barium is useful for diagnosing suspected GI foreign bodies on radiographs.