Learn More
Research Products International Corp Disruption Beads for Tough Tissue, 2.0mm Bead Size, Zirconia Beads, 5.5 g/cc Density, 454 Grams
SDP
Catalog No. 50998371
Encompass
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.
Catalog No. 50-998-371 Supplier Research Products International Corp Supplier No. 9837
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.