Learn More
iQ Biosciences Beagle Canine PBMCs, 10M cells/vial
Catalog No. NC1918325
Encompass_Preferred
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMC) from the beagle canine, a commonly-used dog breed in research. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1918325 Supplier iQ Biosciences Supplier No. IQBCPB102
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMC) from the beagle canine, a commonly-used dog breed in research. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.