Learn More
IPHASE PHARMA SERVICES LLC Beagle Dog Liver Lysosomes,Male,250μL,2mg/ml
Catalog No. NC3860857
Encompass_Preferred
Liver lysosome products have been verified for the activities of cathepsin B, acid phosphatase and nuclease, and can be directly used for in vitro ADC drug linker cleavage and siRNA drug lysosomal escape test verification. The products have undergone strict quality testing, and the test results are accurate, reliable and reproducible.
Catalog No. NC3860857 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 0151C1.01
Liver lysosome products have been verified for the activities of cathepsin B, acid phosphatase and nuclease, and can be directly used for in vitro ADC drug linker cleavage and siRNA drug lysosomal escape test verification. The products have undergone strict quality testing, and the test results are accurate, reliable and reproducible.