Learn More
BRANSON ULTRASONICS CORPORATION (3P) BEAK COVR 2-250ML BEAK M2510
Catalog No. NC3721427
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Beaker Positioning Covers, Fits Model 2800; Configuration: 2 x 250mL, For Use With: 2 x 250mL beaker in ultrasonic benchtop cleaning bath, Helps position beakers securely in bath
Catalog No. NC3721427 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100032527
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Beaker Positioning Covers, Fits Model 2800; Configuration: 2 x 250mL, For Use With: 2 x 250mL beaker in ultrasonic benchtop cleaning bath, Helps position beakers securely in bath