Learn More
Chemglass Life Sciences Chemglass 125 mL Griffin stainless steel beaker (304 SS), low form
SDP
Catalog No. NC3242817
Encompass_Preferred
A 125 mL low-form (Griffin) stainless steel beaker with a seamless, non-corrosive finish and a pour spout for easy liquid transfer.
  • Seamless 304 stainless steel construction for corrosion resistance and easy cleaning.
  • 125 mL capacity suitable for mixing and heating small volumes.
  • Low-form (Griffin) profile for general laboratory use.
  • Pour spout and flared edge for controlled, spill-free pouring.
  • Outer diameter approximately 2-5/8 in and height approximately 2-1/8 in.
  • Durable, easy-to-maintain finish ideal for repeated lab use.
Catalog No. NC3242817 Supplier Chemglass Life Sciences Supplier No. CG1109M125
A 125 mL low-form (Griffin) stainless steel beaker with a seamless, non-corrosive finish and a pour spout for easy liquid transfer.
  • Seamless 304 stainless steel construction for corrosion resistance and easy cleaning.
  • 125 mL capacity suitable for mixing and heating small volumes.
  • Low-form (Griffin) profile for general laboratory use.
  • Pour spout and flared edge for controlled, spill-free pouring.
  • Outer diameter approximately 2-5/8 in and height approximately 2-1/8 in.
  • Durable, easy-to-maintain finish ideal for repeated lab use.