Learn More
Hach Company Beaker 150 mL Glass for Karl Fischer (KF) Titrator, LZE110
Catalog No. NC3368693 Shop All Hach Company Products
Encompass
Glass 150 mL beaker designed for use with Karl Fischer (KF) titrators. Provides a chemically compatible vessel for moisture determinations and replacement of damaged or worn beakers in KF titration workstations.
  • Holds 150 mL volume.
  • Made of glass for chemical compatibility with KF reagents.
  • Designed for use with Karl Fischer titration equipment.
  • Suitable as a replacement or spare laboratory beaker.
  • Easy to clean and maintain for routine titration workflows.
Catalog No. NC3368693 Supplier Hach Company Supplier No. LZE110
Glass 150 mL beaker designed for use with Karl Fischer (KF) titrators. Provides a chemically compatible vessel for moisture determinations and replacement of damaged or worn beakers in KF titration workstations.
  • Holds 150 mL volume.
  • Made of glass for chemical compatibility with KF reagents.
  • Designed for use with Karl Fischer titration equipment.
  • Suitable as a replacement or spare laboratory beaker.
  • Easy to clean and maintain for routine titration workflows.