Learn More
Ace Glass, Inc. Beaker 3L jacketed Ace-Safe hose connections for 1/2in tubing 225mm inside height 133mm inside diameter w/pour spout
SDP
Catalog No. 5000810384
Encompass_Preferred
Beaker 3L jacketed Ace-Safe hose connections for 1/2in tubing 225mm inside height 133mm inside diameter w/pour spout
Catalog No. 50-008-10384 Supplier Ace Glass, Inc. Supplier No. 5341125
Beaker 3L jacketed Ace-Safe hose connections for 1/2in tubing 225mm inside height 133mm inside diameter w/pour spout