Learn More
Chemglass Life Sciences Beaker, 5L, 316L SS, 9.1" H x 7.09" ID
SDP
Catalog No. 5000980610
Encompass
Beakers are made from 316L stainless steel and available in capacities of 1, 2, 3, and 5 liters. Beakers have solid bar sanitary handle, easy-pouring beaker-spout, and graduation marks. All beakers have a seamless sanitary interior and standard 10 Ra finish. Features: Beakers have solid bar sanitary handle. Easy-pouring beaker-spout. Graduation Subdivision lines/marks (without numbers). 316L Polished 10 Ra Finish.
Catalog No. 50-009-80610 Supplier Chemglass Life Sciences Supplier No. CG1109X5000
Beakers are made from 316L stainless steel and available in capacities of 1, 2, 3, and 5 liters. Beakers have solid bar sanitary handle, easy-pouring beaker-spout, and graduation marks. All beakers have a seamless sanitary interior and standard 10 Ra finish. Features: Beakers have solid bar sanitary handle. Easy-pouring beaker-spout. Graduation Subdivision lines/marks (without numbers). 316L Polished 10 Ra Finish.