Learn More
Cole-Parmer Essentials Tri-Corner Beaker, PP, 100 mL, 100/pk
Supplier:  Cole-Parmer 0602003
Encompass_Preferred
Polypropylene (PP) beakers feature a tri-corner design that provides three dripless pouring spouts. Beakers are perfect for use with stirrers — flat bottom keeps stir bars centered. Beakers can withstand temperatures to 250°F (121°C).
Catalog No. NC2000101