Learn More
BUCHI Beaker Flask Gasket
SDP
Catalog No. 05405023 Shop All BUCHI Products
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
05-405-023 PTFE
1 options
Catalog No. 05-405-023 Supplier BUCHI Supplier No. 034152
Add to Cart
Add to Cart
PTFE-coated gasket is used to seal the beaker flask.
  • For use with 1.5 L and 4 L beaker flasks
  • Includes O-ring
Specifications
Material PTFE
Type Gasket
For Use With (Application) Sealing a Beaker Flask
For Use With (Equipment) Beaker Flasks (1.5 L, and 4 L)
Includes O-Ring
Material (Coating) PTFE