Learn More
Andwin Scientific BEAKER GRADUATED 32OZ EA
SDP
Catalog No. 501305867
Encompass_Preferred
This beaker by Polarware is made of medium gauge 304 stainless steel, has a volume capacity of 1000 mL or 32 oz, and has mL and oz graduations. This product is able to withstand temperatures from -450F to 800F. It also has handles that allow easy grip handles.
Catalog No. 50-130-5867 Supplier Andwin Scientific Supplier No. 722200A
This beaker by Polarware is made of medium gauge 304 stainless steel, has a volume capacity of 1000 mL or 32 oz, and has mL and oz graduations. This product is able to withstand temperatures from -450F to 800F. It also has handles that allow easy grip handles.