Learn More
Accuflow Systems Saint-Gobain Chemware Beaker, PTFE, Griffin Low Form, 250 mL
Catalog No. NC2327703
Encompass_Preferred
PTFE Griffin low-form beaker with 250 mL capacity, designed for use with highly corrosive reagents and other demanding laboratory applications. Constructed from PTFE for exceptional chemical resistance and low contamination risk, this reusable beaker is suitable for mixing, heating, and temporary storage of aggressive chemicals.
  • Material: PTFE (polytetrafluoroethylene).
  • Capacity: 250 mL.
  • Form: Griffin low form.
  • Chemical resistance: inert to most corrosive reagents.
  • Color: opaque white.
  • Typical applications: mixing, heating, and storage of aggressive chemicals.
Catalog No. NC2327703 Supplier Accuflow Systems Supplier No. D1069038
PTFE Griffin low-form beaker with 250 mL capacity, designed for use with highly corrosive reagents and other demanding laboratory applications. Constructed from PTFE for exceptional chemical resistance and low contamination risk, this reusable beaker is suitable for mixing, heating, and temporary storage of aggressive chemicals.
  • Material: PTFE (polytetrafluoroethylene).
  • Capacity: 250 mL.
  • Form: Griffin low form.
  • Chemical resistance: inert to most corrosive reagents.
  • Color: opaque white.
  • Typical applications: mixing, heating, and storage of aggressive chemicals.