Learn More
Foxx Life Sciences Borosil Low-Form Griffin Beakers with Pouring Spouts, 100mL, 40/CS
Catalog No. 501542790
Encompass_Preferred
Borosil Low-Form Griffin Beakers are designed with pouring spouts for precise pouring. Uniform wall thickness & stress-free glass make them ideal for heating or cooling. High chemical resistance means Borosil Beakers can be used in nearly any application. Avail. in sizes from 5mL to 10L. Borosil Beakers comply with ISO 3819 & DIN 12331. 500mL & 10L are not covered in ISO or DIN standards. 5mL & 10mL sizes are not graduated. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-154-2790 Supplier Foxx Life Sciences Supplier No. 1000D16
Borosil Low-Form Griffin Beakers are designed with pouring spouts for precise pouring. Uniform wall thickness & stress-free glass make them ideal for heating or cooling. High chemical resistance means Borosil Beakers can be used in nearly any application. Avail. in sizes from 5mL to 10L. Borosil Beakers comply with ISO 3819 & DIN 12331. 500mL & 10L are not covered in ISO or DIN standards. 5mL & 10mL sizes are not graduated. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.