Learn More
Foxx Life Sciences Borosil® Beakers, Griffin Low Form with Spout, 3L (3000mL), 4/CS
Catalog No. 501720204
Encompass_Preferred
Borosil® Low-Form Griffin Beakers are designed with pouring spouts for precise pouring. Uniform wall thickness & stress-free glass make them ideal for heating or cooling. High chemical resistance means Borosil® Beakers can be used in nearly any application. Avail. in sizes from 5mL to 10L. Borosil® Beakers comply with ISO 3819 & DIN 12331. 500mL & 10L are not covered in ISO or DIN standards. 5mL & 10mL sizes are not graduated. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0204 Supplier Foxx Life Sciences Supplier No. 1000D31
Borosil® Low-Form Griffin Beakers are designed with pouring spouts for precise pouring. Uniform wall thickness & stress-free glass make them ideal for heating or cooling. High chemical resistance means Borosil® Beakers can be used in nearly any application. Avail. in sizes from 5mL to 10L. Borosil® Beakers comply with ISO 3819 & DIN 12331. 500mL & 10L are not covered in ISO or DIN standards. 5mL & 10mL sizes are not graduated. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.