Learn More
Chemglass Life Sciences Beaker, Griffin, 2000mL, Polypropylene, Graduated
SDP
Catalog No. 50872718
Encompass_Preferred
Beaker, Griffin, 2000mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.
Catalog No. 50-872-718 Supplier Chemglass Life Sciences Supplier No. CG110817
Beaker, Griffin, 2000mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.