Learn More
Supertek Beaker with Handle (Measuring Jug)
SCI_ED
Catalog No. S01878 Shop All Supertek Scientific Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01878 500 mL
S01877 250 mL
S01875 1000 mL
S01876 2000 mL
S01873 3000 mL
S01874 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S01878 Supplier Supertek Supplier No. CH10165/1
Only null left
Add to Cart
Add to Cart
Polypropylene Beaker with Handle (Measuring Jug) that is autoclavable and has a non-drip spout. These jugs have raised graduations with contact clarity, and are provided with a thumb grip on the handle.
Sturdy polypropylene construction makes these beakers durable, with excellent chemical resistance.
  • Polypropylene
  • Autoclavable
  • Raised graduations
  • Large, strong handles with thumb grip for safe handling of acids, alkalis, and other hazardous chemicals.
Specifications
Capacity (Metric) 500 mL
Material Polypropylene
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Embossed Scale
Style Handled