Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100B TM616
Encompass_Preferred
Aluminum housed mantle for 2L griffin beaker, to 400 degrees C maximum internal operating temperature, 550W, 115V CSA
Catalog No. NC1689316