Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0209072
Encompass_Preferred
Aluminum housed heating mantle for 1L griffin beaker to 400 degrees C maximum internal operating temperature. 430W, 115V. CSA
Catalog No. NC0209072 Supplier Glas Col LLC Supplier No. 100B TM614
Aluminum housed heating mantle for 1L griffin beaker to 400 degrees C maximum internal operating temperature. 430W, 115V. CSA