Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1689317
Encompass_Preferred
Aluminum housed mantle for 4L griffin beaker, to 400 degrees C maximum internal operating temperature, 710W, 115V
Catalog No. NC1689317 Supplier Glas Col LLC Supplier No. 100B TM620
Aluminum housed mantle for 4L griffin beaker, to 400 degrees C maximum internal operating temperature, 710W, 115V