Learn More
Bellco Glass, Inc. Beaker-Heavy Wall,1L
SDP
Catalog No. 501945690
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 1LDimensions: 110 x 145mmCase Quantity: 3
Catalog No. 50-194-5690 Supplier Bellco Glass, Inc. Supplier No. 562001000
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 1LDimensions: 110 x 145mmCase Quantity: 3