Learn More
Bellco Glass, Inc. Beaker-Heavy Wall,2L
SDP
Catalog No. 501945691
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2LDimensions: 134 x 190mmCase Quantity: 3
Catalog No. 50-194-5691 Supplier Bellco Glass, Inc. Supplier No. 562002000
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2LDimensions: 134 x 190mmCase Quantity: 3