Learn More
Bellco Glass, Inc. Beaker-Heavy Wall 50 mL
SDP
Catalog No. 501945685
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 50mLDimensions: 41 x 56mmCase Quantity: 12
Catalog No. 50-194-5685 Supplier Bellco Glass, Inc. Supplier No. 562000050
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 50mLDimensions: 41 x 56mmCase Quantity: 12