Learn More
Bellco Glass, Inc. Beaker-Heavy Wall,4L
SDP
Catalog No. 501945692
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 4LDimensions: 160 x 250mmCase Quantity: 3
Catalog No. 50-194-5692 Supplier Bellco Glass, Inc. Supplier No. 562004000
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 4LDimensions: 160 x 250mmCase Quantity: 3