Learn More
Bellco Glass, Inc. Beaker-Heavy Wall, 5mL
SDP
Catalog No. 501945684
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 5mLDimensions: 20 x 23mmCase Quantity: 12
Catalog No. 50-194-5684 Supplier Bellco Glass, Inc. Supplier No. 562000005
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 5mLDimensions: 20 x 23mmCase Quantity: 12